Skip to Content
Merck
All Photos(1)

Documents

539508

Sigma-Aldrich

PP2A₁, Bovine Kidney

Synonym(s):

Protein Phosphatase 2A₁

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥750 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−20°C

General description

Native, PP2A1 from bovine kidney. A divalent cation-independent protein serine/threonine phosphatase involved in the control of numerous cellular processes. Has a catalytic subunit of 36 kDa, a subunit of 55 kDa, and a regulatory subunit of 60 kDa. Reported to associate with microtubules and to bind to purified tubulin polymerized by TAXOL® Drug.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will release 1.0 nmol of [³²P]Pi from ³²P-labeled phosphorylase per min at 30°C, pH 7.0.

Physical form

In 50 mM Tris-HCl, 14 mM β-ME, 1 mM benzamidine, 1 mM EDTA, 100 µM PMSF, 50% glycerol, pH 7.0.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Hiraga, A. and Tamura, S. 2000. Biochem. J.346, 433.
Ferrigno, P., et al. 1993. Mol. Biol. Cell. 4, 669.
Guo, H. and Damuni, Z. 1993. Proc. Natl. Acad. Sci. USA90, 2500.
Hendrix, P., et al. 1993. J. Biol. Chem.268, 15267.
Amick, G.D., et al. 1992. Biochem. J. 287, 1019.
Cohen, P., et al. 1990. Trends Biochem. Sci.15, 98.
Damuni, Z., et al. 1989. J. Biol. Chem.264, 6412.1

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Taxol is a registered trademark of Bristol-Myers Squibb Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service