Skip to Content
Merck
All Photos(1)

Key Documents

5.38776

Sigma-Aldrich

FGFR4 Inhibitor, BLU9931

Synonym(s):

FGFR4 Inhibitor, BLU9931, Fibroblast Growth Factor Receptor 4 Inhibitor, BLU9931

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 233.00

CHF 233.00


Estimated to ship on31 May 2025



Select a Size

Change View
10 MG
CHF 233.00

About This Item

Empirical Formula (Hill Notation):
C26H22Cl2N4O3
Molecular Weight:
509.38
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 233.00


Estimated to ship on31 May 2025


Assay

≥98% (HPLC)

Quality Level

form

solid

potency

3 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A cell-permeable anilinoquinazoline derivative that acts as a highly potent, irreversible, paralog selective inhibitor of fibroblast growth factor receptor 4 kinase (FGFR4; IC50 = 3 nM; kinact/KI = 0.6 x 105 (mol/L) -1s-1). Acts by forming a covalent bond with Cys552 within the ATP-binding pocket of FGFR4. Has minimal effect on FGFR1, 2, and 3 (IC50 = 591 nM, 493 nM, and 150 nM respectively) and does not display any activity towards a panel of 455 other protein kinases. Suppresses FGFR4 signaling and phosphorylation of downstream components in MDA-MB-453 cells. Also shown to be a potent inhibitor of downstream signaling in Hep 3B hepatocellular carcinoma (HCC) cells expressing FGF19 and induces caspase 3 and 7 activity. Inhibits the growth of Hep 3B HCC xenografts in murine model (100 mg/kg, b.i.d; p.o. for 21 days). Displays desirable pharmacokinetic properties with moderate bioavailability (18%) and t1/2 = 2.3 h (~ 10 mg/kg, p.o.).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
FGFR4
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hagel, M., et al. 2015. Cancer Discov.5, 424.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service