Skip to Content
Merck
All Photos(1)

Key Documents

5.34347

Sigma-Aldrich

hCNT2 Inhibitor

Synonym(s):

hCNT2 Inhibitor, 8-(4-Phenylbenzylamino)adenosine, Human Concentrative Nucleoside Transporter 2 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24N6O4
CAS Number:
Molecular Weight:
448.47
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

An aminoadenosine derivative that acts as a potent human concentrative nucleoside transporter 2 inhibitor. Dose-dependently blocks Na+-dependent inosine uptake in COS-7 cells transiently expressing hCNT2 with an IC50 of 0.64 µM. The anti conformer is suggested to be more bioactive than syn.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Primary Target
hCNT2
Reversible: yes
Target IC50: 640 nM in COS-7 cells

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tatani, K., et al. 2015. ACS Med. Chem. Lett.6, 244.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service