Skip to Content
Merck
All Photos(2)

Key Documents

5.32089

Sigma-Aldrich

Dual DYRK/CLK inhibitor, Cpd 23

Synonym(s):

Dual DYRK/CLK inhibitor, Cpd 23, Pre-mRNA Splicing Modulator, DYRK Inhibitor, Cdc2-like Kinase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O2S
CAS Number:
Molecular Weight:
192.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

potency

60 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

SMILES string

O=C(C)C1=CC2=CC(O)=CC=C2S1

General description

A cell permeable 5-hydroxybenzothiophenylethanone that acts as a dual inhibitor of Cdc2-like kinase 1 (Clk1; IC50 = 60 nM) and dual specificity tyrosine phosphorylation-regulated kinases 1A/1B (Dyrk1A/1B; IC50 = 200 and 100 nM, respectively). Also inhibits Clk4 with high potency. Exhibits much reduced inhibitory effect on Haspin ( IC50 = 800 nM) and has much reduced inhibitory effect on other kinases even at higher concentration (~ 5 µM). Causes a complete disappearance of incomplete and alternatively spliced transcripts and is shown to enhance the generation of the mature Clk1 mRNA splicing product (EC50 = 8.9 µM) in cells.
A cell permeable, dual inhibitor of Cdc2-like kinase 1 (Clk1; IC₅₀ = 60 nM) and dual specificity tyrosine phosphorylation-regulated kinases 1A/1B (Dyrk1A/1B; IC₅₀ = 200 and 100 nM, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
DYRK/CLK

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Schmitt, C., et al. 2014. ACS Med. Chem. Lett.5, 963.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service