Skip to Content
Merck
All Photos(1)

Key Documents

5.30758

Sigma-Aldrich

BMX/BTK Inhibitor II, QL47

Synonym(s):

BMX/BTK Inhibitor II, QL47, 1-(1-Acryloylindolin-6-yl)-9-(1-methyl-1H-pyrazol-4-yl)benzo[h][1,6]naphthyridin-2(1H)-one, BMX Inhibitor II, TEC Inhibitor II, Brutonʹs Tyrosine Kinase Inhibitor V, PI 3-K Inhibitor XXI, BTK Inhibitor V, BLK Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H21N5O2
CAS Number:
Molecular Weight:
447.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

yellow

solubility

glacial acetic acid: 2 mg/mL

storage temp.

−20°C

SMILES string

[n]1(ncc(c1)c2cc3c4[n]([c](ccc4cnc3cc2)=O)c5cc6c(cc5)CCN6C(=O)C=C)C

InChI

1S/C27H21N5O2/c1-3-25(33)31-11-10-17-4-7-21(13-24(17)31)32-26(34)9-6-19-14-28-23-8-5-18(12-22(23)27(19)32)20-15-29-30(2)16-20/h3-9,12-16H,1,10-11H2,2H3

InChI key

RTRNJQOBEOISFQ-UHFFFAOYSA-N

General description

A cell-permeable, BMX-IN-1 (Cat. No. 505021 ) family of acrylamide-containing tricyclic quinoline that acts as a dual BMX & BTK inhibitor (IC50 = 6.7 & 6.6 nM, respectively) by targeting ATP-binding site and covalently modifying kinase hinge cysteine (Cys481 in human BTK) with its eletrophilic acrylamide, while inhibiting TEC, BLK, PI 3-K kinase activity only at higher concentrations (IC50 = 195, 366, and 696 nM, respectively) and exhibiting much reduced or no potency against 29 other kinases (IC50 ≥ 1.2 µM). Likewise, potent inhibition is only seen with BTK in an affinity-based selectivity profiling among a panel of 456 kinases. Shown to effectively inhibit anti-IgM-stimulated BTK Y223 autophosphorylation and downstream effector PLCγ2 Y759 phosphorylation (IC50 = 475 & 318 nM, respectively) without affecting the phosphorylation of upstream kinase Syk, nor S6K and Erk from the PI 3-K/mTOR and Raf/Mek/Erk signaling pathway, respectively. More effective than PCI-32765 (ibrutinib) in inducing PARP & Caspase-3 cleavage in Ramos cultures (Effective conc. <5 µM vs. >10 µM; 8 h) and is more potent than AVL-292, BMX-IN-1, CGI-1746, PCI-32765 in antiproliferation activity against 8 B-cell cancer cell lines (GI50 ranges from 120 to 490 nM; 4.9- and 10.8-fold of BMX-IN-1 potency in Ramos and U2932 cultures, respectively). In addition to loss of kinase activity, irreversible inhibitor-modified BTK via Cys481 covalent bond formation is reported to exhibit shortened cellular half-life. Due to poor microsomal stability, QL47 is not recommended for in vivo studies.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
BMX & BTK
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wu, H., et al. 2014. ACS Chem. Biol.9, 1068.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service