Skip to Content
Merck
All Photos(1)

Key Documents

5.30236

Sigma-Aldrich

SKP2 E3 Ligase Inhibitor IV

Synonym(s):

SKP2 E3 Ligase Inhibitor IV, CRL1 SKP2 Inhibitor IV, p21/Cip1/Waf1 Activator IV, p27/Kip1 Activator IV, S-phase Kinase-associated Protein 2 Inhibitor IV; SCF SKP2 Inhibitor IV, (±)-4-((3-(2,2-Dimethyltetrahydro-2H-pyran-4-yl)-4-phenylbutylamino)methyl)-N,N

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 160.00

CHF 160.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
CHF 160.00

About This Item

Empirical Formula (Hill Notation):
C26H38N2O · 2HCl
CAS Number:
Molecular Weight:
467.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 160.00


Estimated to ship on25 May 2025


Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable dimethylanilinomethylamine compound that inhibits SKP2-dependent cellular p27Kip1 ubiquitination activity (10 to 30 µM; 16 h; in MM1.S myeloma and HeLa cultures) by preventing the incorporation of SKP2 into the Skp1-cullin1-F-box/SCF complex. Selectively induces the accumulation of known SCFSKP2 substrates (p21Cip1, p27Kip1, and p57Kip2) in multiple myeloma RPMI 8226 cells, without inducing HSP-27 phosphorylation or affecting cellular levels of Fbw7 and β-TrCP substrates (c-Jun and β-catenin, respectively) seen with proteasome inhibition by Bortezomib (Cat. No. 504314 ) treatment. Shown to exhibit antiproliferation activity against 13 myeloma lines (GI50 in 3 d from 4.2 to 13.2 µM), including melphalan-, doxorubicin- and steroid-resistant clones, via autophage-initiated cell death induction in a caspase-3-independent manner, and exhibit cancer-selective cytotoxicity against primary CD138+ malignant cells vs. CD138- nonneoplastic blood cells from MM patient blood (viability in 24 h with/without 5 µM drug treatment = 1.38%/66.4% for CD138+ vs. 81.3%/94% CD138-).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Skp1/Skp2 interaction
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Chen, Q., et al. 2008. Blood111, 4690.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service