Skip to Content
Merck
All Photos(1)

Documents

529542

Sigma-Aldrich

PSD95 Inhibitor, Tat-N-dimer

The PSD95 Inhibitor, Tat-N-dimer controls the biological activity of PSD95. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Postsynaptic Density Protein-95 Inhibitor, Tat-N-dimer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

potency

4.6 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL
water: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, dimeric pentapeptide (IETDV) derived from GluN2B C-terminal and linked through a polyethylene glycol (PEG)-based linker that inhibits the ternary protein complex of PSD-95 (postsynaptic density protein-95), nNOS, and NMDA by directly binding to the tandem PDZ1 and 2 domains of PSD-95 (Ki = 4.6 nM). This inhibition results in the uncoupling between NMDA receptor activity and nitric oxide production without affecting NMDA-mediated ion channel gating function and pro-survival signaling pathways. Exhibits about 1000-fold higher potency than the monomeric Tat-NR2B9c. Exhibits improved plasma stability, with t1/2 ~ 80 hours. Shown to cross the blood-brain barrier and reduce infarct volume by about 40% and restore motor functions in mice subjected to focal ischemic brain damage.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PSD95
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bach, A., et al. 2012. Proc Natl Acad Sci USA.109 3317.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service