Skip to Content
Merck
All Photos(1)

Key Documents

524572

Sigma-Aldrich

Phosphatase, Alkaline, Calf Intestine

Synonym(s):

Phosphatase, Alkaline, Calf Intestine, Alkaline Phosphatase, CIAP

Sign Into View Organizational & Contract Pricing

Select a Size

1 KU
CHF 77.00
5 KU
CHF 391.00

CHF 77.00


Estimated to ship on01 June 2025



Select a Size

Change View
1 KU
CHF 77.00
5 KU
CHF 391.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

CHF 77.00


Estimated to ship on01 June 2025


biological source

bovine (calf) intestine

Quality Level

form

liquid

specific activity

≥1500 U/mg

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

pI 

5.7

solubility

dilute buffer: soluble
water: soluble

shipped in

ambient

storage temp.

2-8°C

General description

Native, alkaline phosphatase from calf intestine. Homodimeric enzyme that hydrolyzes phosphate esters of primary and secondary alcohols, amines, and phenols. Useful in hydrolysis of 5′-terminal phosphates of DNA and RNA. Note: 1KU = 1,000 units.
Note: 1 KU = 1000 units.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol of pNPP per min at 25°C, pH 9.6.

Physical form

In 6 mM MgCl, 6 mM Tris-HCl, 0.12mM ZnCl, 40% glycerol, pH 7.6.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service