Skip to Content
Merck
All Photos(1)

Key Documents

516531

Sigma-Aldrich

Peroxidase, Horseradish

Synonym(s):

Peroxidase, Horseradish

Sign Into View Organizational & Contract Pricing

Select a Size

5 KU
CHF 87.30
25 KU
CHF 592.00

CHF 87.30


Estimated to ship on31 May 2025



Select a Size

Change View
5 KU
CHF 87.30
25 KU
CHF 592.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

CHF 87.30


Estimated to ship on31 May 2025


form

lyophilized solid

Quality Level

specific activity

≥200 U/mg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

0.1 M phosphate, pH 6.0-7.0: 5 mg/mL

absorbance ratio

RZ3.0

shipped in

ambient

storage temp.

−20°C

SMILES string

[O+H2]O[O-]

InChI

1S/H2O3/c1-3-2/h1-2H

InChI key

JSPLKZUTYZBBKA-UHFFFAOYSA-N

General description

Note: 1 KU = 1000 units.
Suitable for conjugation to proteins and antibodies and as a cytochemical marker in immunohistochemical staining. Has an optimal pH of 7.0.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will catalyze the production of 1 mg purpurogallin in 20 s at 20°C, pH 6.0.

Reconstitution

Following reconstitution, aliquot and refrigerate (4°C). Stock solutions are stable for up to 1 year at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service