Skip to Content
Merck
All Photos(1)

Key Documents

5.06236

Sigma-Aldrich

Tiagabine Hydrochloride

Synonym(s):

Tiagabine Hydrochloride, GABA reuptake inhibitor, Tiagabine, Gabitril, GAT-1 GABA Transporter Inhibitor, Tiagabine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 113.00

CHF 113.00


Estimated to ship on30 May 2025



Select a Size

Change View
10 MG
CHF 113.00

About This Item

Empirical Formula (Hill Notation):
C20H25NO2S2 · xHCl
CAS Number:
Molecular Weight:
375.55 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 113.00


Estimated to ship on30 May 2025


Assay

≥98% (HPLC)

Quality Level

form

solid

potency

67 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

water: soluble

storage temp.

2-8°C

SMILES string

[s]1c(c(cc1)C)C(=CCCN3C[C@@H](CCC3)C(=O)O)c2[s]ccc2C.[Cl-].[H+]

InChI

1S/C20H25NO2S2.ClH/c1-14-7-11-24-18(14)17(19-15(2)8-12-25-19)6-4-10-21-9-3-5-16(13-21)20(22)23;/h6-8,11-12,16H,3-5,9-10,13H2,1-2H3,(H,22,23);1H/t16-;/m1./s1

InChI key

YUKARLAABCGMCN-PKLMIRHRSA-N

General description

A potent uptake inhibitor highly selective for GABA Transport-1 (IC50 = 67 nM in vivo). Clinically used as an anticonvulsant. Often used in studies related to anxiety disorders and neural damages resulted from epileptics.

Biochem/physiol Actions

Primary Target
GAT-1

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Laughlin, T., et al. 2002. J. Pharmacol. Exp. Ther.302, 1168.

Hu, Q., et al. 1997. Brain Res.753,260.
Suzdak, P., et al., 1995. Epilepsia.36, 612.
Andersen, E., et al., 1993. J. Med. Chem.36, 1716.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service