Skip to Content
Merck
All Photos(1)

Documents

5.06073

Sigma-Aldrich

Glutaminase Inhibitor, Compound 968

InSolution, ≥99%, A cell-permeable benzophenanthridinone compound

Synonym(s):

InSolution Glutaminase Inhibitor, Compound 968, GAC Inhibitor, GLS1 Inhibitor I, KGA Inhibitor I, Kidney-Type Glutaminase Inhibitor I, 5-(3-Bromo-4-(dimethylamino)phenyl)-2,2-dimethyl-2,3,5,6-tetrahydrobenzo[a]phenanthridin-4(1H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27BrN2O
Molecular Weight:
475.42
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

General description

A cell-permeable benzophenanthridinone compound that acts as an allosteric and reversible inhibitor of mitochondrial glutaminase activity and represses the growth and invasive activity in glutaminase upregulated fibroblasts and in tumor cells (IC50 ≤ 10 µM in NIH3T3 cells stably expressing Dbl, Cdc42-F28L, Rac-F28L or RhoC-F30L mutants and in SKBR3 and MDA-MB231 cancer cells). Shown to effectively shrink tumor size in P-493 B xenografted mouse model (200 µg per inj. i.p., 12 days).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
mitochondrial glutaminase
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 10 mM (5 mg/1.05 ml) sterile-filtered solution of Glutaminase Inhibitor, Compound 968 (Cat. No. 352010) in DMSO.

Reconstitution

Following initial thaw slight warming may be required to resolubilize. Aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.

Other Notes

Wang, J.B., et al. 2010. Cancer Cell18, 207.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service