Skip to Content
Merck
All Photos(1)

Key Documents

5.06048

Sigma-Aldrich

TAT-Beclin-1

≥97% (HPLC), powder, Beclin-1 activator, Calbiochem®

Synonym(s):

Beclin-1 Activator I, TAT-Beclin-1, Atg6 Activator I, Beclin1-GAPR-1 Interaction Blocker I, Vps30 Activator I, H₂N-YGRKKRRQRRR-GG-TNVFNATFEIWHDGEFGT-CO₂H, Autophagy Inducer IV

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 429.00

CHF 429.00


Estimated to ship on09 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
CHF 429.00

About This Item

Empirical Formula (Hill Notation):
C164H251N57O45
Molecular Weight:
3741.10
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 429.00


Estimated to ship on09 April 2025


Request a Bulk Order

Product Name

Beclin-1 Activator I, TAT-Beclin-1,

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 50 mg/mL
water: 50 mg/mL

storage temp.

2-8°C

General description

Beclin-1 Activator I, TAT-Beclin-1, is a cell-permeable Atg6/Beclin1-derived autophagy-inducing peptide. Activates Beclin1 by competing against its negative regulator GAPR-1/GLIPR2. TAT-Beclin-1 (Minimum effective conc. ≤10 µM assessed by increased LC3-II/LC3-I ratio & p62 downregulation post 3 h treatment in A549, COS-7, HBEC30-KT, HCC827, HeLa, MEF, THP-1 cultures) that is composed of essential HIV-1 virulence factor Nef-binding sequence derived from human Atg6/Beclin1 (aa 267-284) evolutionarily conserved domain (ECD) with substitutions at three non-species-conserved residues (H275E, S279D, and Q280E) for enhanced solubility and N-terminally fused to the membrane-permeant HIV-1 Tat protein transduction domain (PTD) sequence (aa 47-57) via a -Gly-Gly- linkage to facilitate cellular delivery and Beclin1 activation via competitive binding to its negative regulator "Golgi-associated plant pathogenesis-related protein-1" (GAPR-1/GLIPR2) on the Golgi surface. Shown to effectively induce autosis, autophagy-dependent death morphologically and pharmacologically distinct from apoptosis and necrosis, in various cell cultures of human, rat, and murine origin in a dose- and time-dependent manner (Minimum effective conc. >5 µM post 5 h treatment as assessed by HeLa Trypan blue staining; Fold of control Trypan blue-positive population/20 µM peptide exposure time = 3/1 h, 9/4 h, 20/8 h), blockable by autophagy inhibitor 3-MA (10 mM, Cat. no. 189490; [T-B] = 20 µM) or via Na+/K+-ATPase inhibition by cardiac glycosides, but insensitive to Z-VAD-FMK (Cat. no. 219007) or Nec-1 (Cat. nos. 480065 & 505224) treatment. Autophagy induction by Tat-Beclin1 treatment is also reported to result in clearance of small (<1 µm) CFP-htt103Q aggregates in HeLa cells (59% and 85% reduction, respectively, of aggregates-positive population and average aggregates per cell; 20 µM/4h/d for 2 d) and non-cytotoxic autophagy induction with lower level of Tat-Beclin1 dosage and limited exposure time is demonstrated to be therapeutically beneficial in blocking RNA viral replications both in cultures (4 h, 10 µM peptide treatment 4 h post CHIKV, SINV, or WNV infection in HeLa cultures; 0.5-5 µM peptide pre-treatment 24 h prior to HIV infection in human MDM cultures) in vitro and in CHIKV-inoculated neonatal mice (15 mg/kg/day i.p.) in vivo. While Tat-Beclin1 bioavailability is limited to the peripheral tissues in mice, a peptide based on Tat-Beclin1 retro-inverso sequence (Cat. no. 506416) shows efficacy in a murine model of neonatal WNV CNS infection.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GAPR-1/GLIPR2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg~Gly-Gly~Thr-Asn-Val-Phe-Asn-Ala-Thr-Phe-Glu-Ile-Trp-His-Asp-Gly-Glu-Phe-Gly-Thr-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Increased drug potency is reported in acidified (pH 7.0) serum-free cultures.

Legal Information

Sold under license of US Patent 8,722,628.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service