Skip to Content
Merck
All Photos(1)

Key Documents

5.04345

Sigma-Aldrich

BX471

Synonym(s):

BX471, N-[5-chloro-2-[2-[(2R)-4-[(4-fluorophenyl)methyl]-2-methyl-1-piperazinyl]-2-oxoethoxy]phenyl]-urea, ZK-811752

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24ClFN4O3
CAS Number:
Molecular Weight:
434.89
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

1-5 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to beige

solubility

DMSO: ≥25 mg/mL

storage temp.

2-8°C

InChI

1S/C21H24ClFN4O3/c1-14-11-26(12-15-2-5-17(23)6-3-15)8-9-27(14)20(28)13-30-19-7-4-16(22)10-18(19)25-21(24)29/h2-7,10,14H,8-9,11-13H2,1H3,(H3,24,25,29)/t14-/m1/s1

InChI key

XQYASZNUFDVMFH-CQSZACIVSA-N

General description

An orally active, selective antagonist for chemokine receptor 1 (CCR-1) (Ki = 1-5 nM). Has been used in studies related to intracellular calcium mobilization and migration of leukocytes mediated through CCR-1.

Biochem/physiol Actions

Primary Target
CCR1

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Shahlaei, M., et al. 2011. J. Chemical Information and Modeling.51, 2717.

Reuss, R., et al. 2010. Mult Scler.16, 366.

Gladue, R., et al. 2010. Curr Top Med Chem10, 1268.

Vandemeulebroecke; M., et al. 2009. British J. of Clinical Pharmacol.68, 435.

Richard Horuk, R. 2005. Mini-Reviews in Medicinal Chemistry.5, 791.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service