5.00859
Vitamin E
dry powder, (50 g all-rac-a-tocopheryl acetate in 100 g subst), EMPROVE® ESSENTIAL, Ph. Eur., BP, USP
Pharma Manufacturing
Synonym(s):
(±)-α-Tocopherol, Vitamin E dry powder (50 g all-rac-α-tocopheryl acetate in 100 g substance), DL-all-rac-α-Tocopherol, Vitamin E
About This Item
Recommended Products
Agency
BP
Ph. Eur.
USP
Quality Level
product line
EMPROVE® ESSENTIAL
form
dry powder
solid
pH
7 (20 °C, 50 g/L in H2O, slurry)
density
0.950 g/mL at 20 °C (lit.)
application(s)
pharmaceutical
storage temp.
2-25°C
SMILES string
CC(C)CCCC(C)CCCC(C)CCCC1(C)CCc2c(C)c(O)c(C)c(C)c2O1
InChI
1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3
InChI key
GVJHHUAWPYXKBD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service