Skip to Content
Merck
All Photos(1)

Key Documents

5.00469

Sigma-Aldrich

S107

≥98% (HPLC), solid, RyR-Calstabin interaction stabilizer, Calbiochem®

Synonym(s):

RyR-Calstabin Interaction Stabilizer, S107, 7-methoxy-4-methyl-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine, Ryanodine Receptor Stabilizer, Rycal S107

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 281.00

CHF 281.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
CHF 281.00

About This Item

Empirical Formula (Hill Notation):
C11H15NOS
CAS Number:
Molecular Weight:
209.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 281.00


Estimated to ship on20 May 2025


Request a Bulk Order

Product Name

RyR-Calstabin Interaction Stabilizer, S107,

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

S1CCN(Cc2c1ccc(c2)OC)C

InChI key

BGVCEGVSQDOGSB-UHFFFAOYSA-N

General description

RyR-Calstabin Interaction Stabilizer, S107 can protect hippocampal neurons from GABAergic synaptic changes caused by Abeta42 oligomers.[1]

Application

RyR-Calstabin Interaction Stabilizer, S107 has been used in culture media for hypoxic exposures to study the impact of hypoxia on sarcoplasmic reticulum calcium leakage and expression of ryanodine receptor2 (RyR2).[2]

Biochem/physiol Actions

Primary Target
RyR2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ryanodine receptor-calstabin interaction stabilizer S107 protects hippocampal neurons from GABAergic synaptic alterations induced by Abeta42 oligomers
Hidisoglu E, et al.
The Journal of Physiology, 600(24), 5295-5309 (2022)
Hypoxia-Induced Sarcoplasmic Reticulum Ca2+ Leak Is Reversed by Ryanodine Receptor Stabilizer JTV-519 in HL-1 Cardiomyocytes
Trinh MD, et al.
Anatolian journal of cardiology, 26(6), 476-476 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service