Skip to Content
Merck
All Photos(1)

Key Documents

491207

Sigma-Aldrich

Novobiocin sodium salt

≥90% (HPLC), powder, DNA gyrase inhibitor, Calbiochem®

Synonym(s):

Novobiocin, Sodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H35N2O11 · Na
CAS Number:
Molecular Weight:
634.61
MDL number:
UNSPSC Code:
51286504
Pricing and availability is not currently available.

Product Name

Novobiocin, Sodium Salt,

Assay

≥90% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to yellow

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].N([C-]2C(=O)Oc3c(ccc(c3C)OC4OC(C(C(C4O)OC(=O)N)OC)(C)C)C2=O)C(=O)c1cc(c(cc1)O)CC=C(C)C

InChI

1S/C31H35N2O11.Na/c1-14(2)7-8-16-13-17(9-11-19(16)34)27(37)33-21-22(35)18-10-12-20(15(3)24(18)42-28(21)38)41-29-23(36)25(43-30(32)39)26(40-6)31(4,5)44-29;/h7,9-13,23,25-26,29,34,36H,8H2,1-6H3,(H2,32,39)(H,33,37);/q-1;+1

InChI key

AXOUUAINTJNFRS-UHFFFAOYSA-N

General description

A DNA gyrase inhibitor that can be used for the production of positively supercoiled plasma DNA. Targets the nucleotide-binding site of gyrase B. Exhibits antibacterial properties. Inhibits retrovirus RNA-dependent DNA polymerases. Inhibits LPS-induced production of pro-inflammatory cytokines. Reported to act as a potent inhibitor of ADP ribosylation. Also inhibits protein synthesis and alters the phosphorylation state of several cytosolic proteins.
DNA gyrase inhibitor used for the production of positively supercoiled plasma DNA. Inhibits retrovirus RNA-dependent DNA polymerases. Also a potent inhibitor of ADP ribosylation eliminates the wild-type pMG110 plasmid from E. coli.

Biochem/physiol Actions

Cell permeable: no
Primary Target
retrovirus RNA-dependent DNA polymerases
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for 3 months at 4°C.

Other Notes

Marcu, M.G., et al. 2000. J. Natl. Cancer Inst.92, 242.
Luhrmann, A., et al. 1998. Antimicrob. Agents Chemotherp.42, 1911.
Hooper, D.C., et al. 1984. Antimicrob. Agents Chemother. 25, 586.
Lockshon, D., and Morris, D.R. 1983. Nucleic Acid. Res. 11, 2999.
Sumiyoshi, Y., et al. 1983. J. Gen. Virol. 64, 2329.
Staudenbauer, W.L. 1975. J. Mol. Biol. 96, 201.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service