Skip to Content
Merck
All Photos(1)

Key Documents

482777

Sigma-Aldrich

Nitric Oxide Synthase, Inducible, Mouse, Recombinant, E. coli

Synonym(s):

Inducible Nitric Oxide Synthase, iNOS, Mouse, Recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

50 U
CHF 548.00

CHF 548.00


Estimated to ship on14 April 2025



Select a Size

Change View
50 U
CHF 548.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

CHF 548.00


Estimated to ship on14 April 2025


recombinant

expressed in E. coli

Quality Level

form

liquid

specific activity

≥2.9 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, mouse inducible NOS expression in E. coli.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Harmful (C)

Unit Definition

One unit is defined as the amount of enzyme that will produce 1.0 nmol nitric oxide per min at 37°C, pH 7.4.

Physical form

In 50 mM HEPES, 10% glycerol, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Stevens-Truss, R., and Marletta, M.A. 1995. Biochemistry 34, 15638.
Hevel, J.M., and Marletta, M.A. 1994. Methods Enzymol. 233, 250.
Marletta, M.A. 1994. J. Med. Chem. 37, 1899.
White, K.A., and Marletta, M.A. 1992. Biochemistry 31, 6627.
Hevel, J.M., et al. 1991. J. Biol. Chem. 266, 22789.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service