Skip to Content
Merck
All Photos(1)

Key Documents

481990

Sigma-Aldrich

Nigericin, Sodium Salt, Streptomyces hygroscopicus

Disrupts the membrane potential and stimulates ATPase activity in mitochondria.

Synonym(s):

Nigericin, Sodium Salt, Streptomyces hygroscopicus, Antibiotic K178

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H67O11 · Na
CAS Number:
Molecular Weight:
746.94
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

chloroform: soluble
ethanol: soluble
ethyl acetate: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C40H68O11.Na/c1-21-11-12-28(46-33(21)26(6)36(42)43)17-29-18-30(45-10)27(7)40(48-29)25(5)19-38(9,51-40)32-13-14-37(8,49-32)35-23(3)16-31(47-35)34-22(2)15-24(4)39(44,20-41)50-34;/h21-35,41,44H,11-20H2,1-10H3,(H,42,43);/q;+1/p-1/t21-,22-,23-,24+,25?,26?,27+,28+,29+,30+,31+,32?,33?,34-,35?,37?,38?,39-,40+;/m0./s1

InChI key

MOYOTUKECQMGHE-KKCUGXASSA-M

General description

A monocarboxylate that catalyzes the electroneutral exchange of an alkali metal for H+ (antiport). Transports monovalent cations across membranes with the following specificities: K+>Rb+>Cs+>>Na+. An antibiotic that kills bacteria by facilitating the diffusion of ions across membranes. Disrupts the membrane potential and stimulates mitochondrial ATPase activity.
Disrupts the membrane potential and stimulates ATPase activity in mitochondria. Transports monovalent cations across membranes with the following specificity: K+>Rb+>Cs+>>Na+.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Disrupts the membrane potential and stimulates ATPase activity in mitochondria.
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze. Stock solutions are stable for 1 month at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Moriyama, Y., et al. 1994. J. Biochem.115, 213.
Eytan, G.D., et al. 1990. J. Biol. Chem.265, 12949.
Alonso, M.A., and Carrasco, L. 1981. FEBS Lett.127, 112.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandip Ghosh et al.
Biosensors & bioelectronics, 155, 112115-112115 (2020-03-29)
Fluorescence based intracellular pH nanoprobes have been developed that overcomes the limitations imposed by shallow penetration depth of ultraviolet excitation, photostability, phototoxicity, and interference from background autofluorescence. In this study, we have constructed a Förster Resonance Energy Transfer (FRET) based
Liang Tao et al.
Cell death and differentiation, 28(4), 1418-1433 (2020-11-20)
Hepatocyte cell death and liver inflammation have been well recognized as central characteristics of nonalcoholic steatohepatitis (NASH), however, the underlying molecular basis remains elusive. The kinase receptor-interacting protein 1 (RIP1) is a multitasking molecule with distinct functions in regulating apoptosis
Juan Ma et al.
The EMBO journal, 40(18), e108249-e108249 (2021-07-24)
SARS-CoV-2 is an emerging coronavirus that causes dysfunctions in multiple human cells and tissues. Studies have looked at the entry of SARS-CoV-2 into host cells mediated by the viral spike protein and human receptor ACE2. However, less is known about
Pan Pan et al.
Nature communications, 12(1), 4664-4664 (2021-08-04)
Excessive inflammatory responses induced upon SARS-CoV-2 infection are associated with severe symptoms of COVID-19. Inflammasomes activated in response to SARS-CoV-2 infection are also associated with COVID-19 severity. Here, we show a distinct mechanism by which SARS-CoV-2 N protein promotes NLRP3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service