Skip to Content
Merck
All Photos(1)

Key Documents

481480

Sigma-Aldrich

NF-κB SN50

≥97% (HPLC), lyophilized, NF-κB translocation inhibitor, Calbiochem®

Synonym(s):

NF-κB SN50, Cell-Permeable Inhibitor Peptide

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
CHF 260.00
1 MG
CHF 391.00

CHF 260.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
500 μG
CHF 260.00
1 MG
CHF 391.00

About This Item

Empirical Formula (Hill Notation):
C129H230N36O29S
Molecular Weight:
2781.50
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 260.00


Estimated to ship on28 May 2025


Request a Bulk Order

Product Name

NF-κB SN50, Cell-Permeable Inhibitor Peptide, NF-κB SN50, CAS 213546-43-3, is a cell-permeable peptide that contains the NLS sequence of NF-κB p50 linked to the hydrophobic region of K-FGF. Inhibits translocation of the NF-κB into the nucleus.

Quality Level

Assay

≥97% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Contains the nuclear localization sequence (NLS) of the transcription factor NF-κB p50 linked to the hydrophobic region (h-region) of the signal peptide of Kaposi fibroblast growth factor (K-FGF). The N-terminal K-FGF h-region confers cell-permeability, while the NLS (360-369) inhibits translocation of the NF-κB active complex into the nucleus. In murine endothelial LE-II cells induced by LPS, NF-κB nuclear translocation is maximally inhibited at 18 µM.
NF-κB SN50, CAS 213546-43-3, is a cell-permeable peptide that contains the NLS sequence of NF-κB p50 linked to the hydrophobic region of K-FGF. Inhibits translocation of the NF-κB into the nucleus.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
translocation of the NF-κB active complex into the nucleus
Product does not compete with ATP.

Warning

Toxicity: Standard Handling (A)

Sequence

H-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Val-Gln-Arg-Lys-Arg-Gln-Lys-Leu-Met-Pro-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Avoid freeze/thaw cycles of solutions.

Other Notes

Lin, Y.-Z., et al. 1995. J. Biol. Chem. 270, 14255.

Legal Information

Sold under license of U.S. Patents 5,807,746 and 6,043,339 for research use only. Licenses for commercial manufacture, commercial use, diagnostics, or therapeutics may be obtained by contacting Vanderbilt University, Nashville, Tennessee.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna R Reed et al.
PloS one, 7(9), e45877-e45877 (2012-10-03)
Tumor formation is an extensive process requiring complex interactions that involve both tumor cell-intrinsic pathways and soluble mediators within the microenvironment. Tumor cells exploit the intrinsic functions of many soluble molecules, including chemokines and their receptors, to regulate pro-tumorigenic phenotypes
Taewoo Yang et al.
Journal of experimental & clinical cancer research : CR, 41(1), 82-82 (2022-03-05)
BCR-ABL-independent drug resistance is a barrier to curative treatment of chronic myeloid leukemia (CML). However, the molecular pathways underlying BCR-ABL-independent tyrosine kinase inhibitor (TKI) resistance remain unclear. In silico bioinformatic analysis was performed to identify the most active transcription factor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service