Skip to Content
Merck
All Photos(1)

Key Documents

480418

Sigma-Aldrich

NF279

A suramin analog that acts as a highly selective, competitive, and reversible ATP-antagonist of P2X receptor (IC₅₀/KB ~ 1 µM in smooth muscle).

Synonym(s):

NF279, 8,8ʹ-(Carbonylbis(imino-4,1-phenylenecarbonylimino-4,1-phenylenecarbonylimino))bis-1,3,5-naphthalenetrisulfonic Acid, Na

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 192.00

CHF 192.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CHF 192.00

About This Item

Empirical Formula (Hill Notation):
C49H30N6O23S6 · 6Na
CAS Number:
Molecular Weight:
1401.12
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 192.00


Estimated to ship on22 May 2025


Request a Bulk Order

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

DMSO: 14 mg/mL
water: 35 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[S](=O)(=O)([O-])c1c2c(c(cc(c2)[S](=O)(=O)[O-])[S](=O)(=O)O)c(cc1)\N=C(/[O-])\c3ccc(cc3)NC(=O)c4ccc(cc4)NC(=O)Nc5ccc(cc5)C(=O)Nc6ccc(cc6)\C(=N\c7c8c(c(cc7)[S](=O)(=O)[O-])cc(cc8[S](=O)(=O)O)[S](=O)(=O)[O-])\[O-]

InChI

1S/C49H36N6O23S6.6Na/c56-45(50-29-9-1-27(2-10-29)47(58)54-37-17-19-39(81(67,68)69)35-21-33(79(61,62)63)23-41(43(35)37)83(73,74)75)25-5-13-31(14-6-25)52-49(60)53-32-15-7-26(8-16-32)46(57)51-30-11-3-28(4-12-30)48(59)55-38-18-20-40(82(70,71)72)36-22-34(80(64,65)66)24-42(44(36)38)84(76,77)78;;;;;;/h1-24H,(H,50,56)(H,51,57)(H,54,58)(H,55,59)(H2,52,53,60)(H,61,62,63)(H,64,65,66)(H,67,68,69)(H,70,71,72)(H,73,74,75)(H,76,77,78);;;;;;/q;6*+1/p-6

InChI key

RJMCMLNRWDKUDB-UHFFFAOYSA-H

General description

A suramin analog that acts as a highly selective, competitive, and reversible ATP-antagonist of P2X receptor (IC50/KB ~1 µM in smooth muscle). Effectively discriminates between P2Y and P2X receptors with no discernible effects on α1A adrenoceptors, adenosine A1 and A2B receptors, histamine H1, muscarinic M3 and neuronal nicotinic acetylcholine receptors. Displays a selectivity profile of P2X1 >P2X2 >P2X3 >P2X4 (IC50 = 19 nM, 770 nM, 1.62 µM and >300 µM, respectively) in Xenopus oocytes pre-incubated with ATP. In rat and human tissues, exhibits a potency profile of rat P2X1 > human P2X1 » rat P2X2 > rat P2X3 ~human P2X7 » human P2X4. Not degraded by ecto-nucleotidases.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
P2X receptor
Product competes with ATP.
Reversible: yes
Target IC50: ~ 1 µM as antagonist of P2X receptor in smooth muscle; 19 nM, 770 nM, 1.62 µM and <300 µM, against P2X1, P2X2, P2X3, P2X4, respectively, in Xenopus oocytes pre-incubated with ATP

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Klapperstück, M., et al. 2000. Eur. J. Pharmacol.387, 245.
Rettinger, J., et al. 2000. Neuropharmacol.39, 2044.
Lambrecht, G., et al. 1999. Prog. Brain Res.120,107.
Damer, S., et al. 1998. Eur. J. Pharmacol.350, R5.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service