Skip to Content
Merck
All Photos(1)

Key Documents

480012

Millipore

NDSB Set

Contains the following non-detergent sulfobetaines: 5 g of NDSB-195, 25 g of NDSB-201, and 5 g of NDSB-256.

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
Inquire


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 SET
Inquire

About This Item

UNSPSC Code:
12352200
NACRES:
NA.21


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

15-25°C

General description

Contains the following non-detergent sulfobetaines: 5 g of NDSB-195 (Cat. No. 480001), 25 g of NDSB-201 (Cat. No. 480005), and 5 g of NDSB-256 (Cat. No. 480010). Supplied with a data sheet.
Non-detergent sulfobetaines (NDSBs) are zwitterionic compounds. Like zwittergents (zwitterionic detergents), NDSBs carry the sulfobetaine hydrophilic head group. However, unlike zwittergents, the hydrophobic group in NDSBs is too short for micellar formation even at concentrations as high as 1M. Hence, NDSBs do not behave like detergents. Because of their zwitterionic nature, NDSBs were first employed to neutralize electrostatic interactions without increasing the conductivity in native isoelectrofocusing gels. Recently they have found utility in various other applications including isolation of membrane proteins, purification of nuclear proteins and halophilic proteins, renaturation of chemically and thermally denatured proteins and protein crystallization. They are potentially useful in solubilization and renaturation of fusion proteins from inclusion bodies. It is hypothesized that the hydrophobic group, although short, interacts with the hydrophobic regions of the protein preventing aggregation during renaturation. Also, presumably, the contribution from the short hydrophobic groups combined with charge neutralization ability of the sulfobetaine group results in higher yields of membrane proteins.



NDSBs do not interfere with enzymatic assays based on chromogenic substrates bearing nitrophenyl groups and do not inhibit the activities of β-galactosidase and alkaline phosphatase, the two enzymes tested thus far. NDSB-195 has the added advantage in that it does not absorb at 280 nm, making it compatible with protein purification procedures in which the protein concentrations are determined by absorbance measurements at 280 nm. Supplied with a data sheet.

Warning

Toxicity: Irritant (B)

Reconstitution

Store lyophilized products at room temperature. Following reconstitution, store in the refrigerator (4°C) or at room temperature (20°C).

Other Notes

Goldberg, M.E., et al. 1996. Folding and Design1, 21.
Goldberg, M.E., et al. 1995. Folding & Design1, 21.
Vuillard, L., et al. 1995. Biochem. J.305, 337.
Vuillard, L., et al. 1995. Anal. Biochem.230, 290.
Vuillard, L., et al. 1994. FEBS Lett.353, 294.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service