Skip to Content
Merck
All Photos(1)

Key Documents

475820

Millipore

Mitomycin C, Streptomyces caespitosus, Carrier-Free

Antibiotic and carcinostatic agent. Inhibits DNA synthesis by cross-linking DNA at guanine and adenine residues; disrupts base pairing. Induces apoptosis in gastric cancer cells.

Synonym(s):

Mitomycin C, Streptomyces caespitosus, Carrier-Free, Mitocin C, MMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N4O5
CAS Number:
Molecular Weight:
334.33
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

purple

solubility

water: <1 mg/mL
methanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C15H18N4O5/c1-5-9(16)12(21)8-6(4-24-14(17)22)15(23-2)13-7(18-13)3-19(15)10(8)11(5)20/h6-7,13,18H,3-4,16H2,1-2H3,(H2,17,22)/t6-,7+,13+,15-/m1/s1

InChI key

NWIBSHFKIJFRCO-WUDYKRTCSA-N

General description

Antibiotic and carcinostatic agent. Inhibits DNA synthesis by cross-linking DNA at guanine and adenine residues; disrupts base pairing. Induces apoptosis in gastic cancer cells. Blocks the cell cycle at the G2 phase.
Antibiotic and carcinostatic agent. Inhibits DNA synthesis by cross-linking DNA at guanine and adenine residues; disrupts base pairing. Induces apoptosis in gastric cancer cells. Blocks the cell cycle at the G2 phase. Potency: ≥970 µg/mg.

Biochem/physiol Actions

Potency: ≥ 970 μg/mg mitomycin C

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Kryt, F.A., et al. 1996. Blood 89, 938.
Schwartz, G.K., et al. 1995. J. Natl. Cancer Inst. 87, 1394.
Folnan, R.S., 1993. Oncology 50, 24.
Tomasz, M., et al. 1987. Science 235, 1204.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fangzhou Lou et al.
British journal of pharmacology, 181(8), 1290-1307 (2023-09-26)
Psoriasis vulgaris is a refractory skin inflammatory disorder with 80% of the cases belonging to the mild-to-moderate type, which can be controlled by topical treatment. Nevertheless, the drugs for external use have not been upgraded for decades. We modified acetyl-11-keto-beta-boswellic
Arvind Panday et al.
Molecular cell, 81(11), 2428-2444 (2021-04-22)
Repair pathway "choice" at stalled mammalian replication forks is an important determinant of genome stability; however, the underlying mechanisms are poorly understood. FANCM encodes a multi-domain scaffolding and motor protein that interacts with several distinct repair protein complexes at stalled
Yuchuan Li et al.
Cancer cell international, 23(1), 276-276 (2023-11-18)
Despite therapeutic advances, the prognosis of pancreatic ductal adenocarcinoma (PDAC) remains extremely poor. Metabolic reprogramming is increasingly recognized as a key contributor to tumor progression and therapy resistance in PDAC. One of the main metabolic changes essential for tumor growth
Romie Angelo G Azur et al.
PloS one, 19(5), e0300413-e0300413 (2024-05-13)
Castration-resistant prostate cancer (CRPC) is associated with resistance to androgen deprivation therapy, and an increase in the population of neuroendocrine (NE) differentiated cells. It is hypothesized that NE differentiated cells secrete neuropeptides that support androgen-independent tumor growth and induce aggressiveness

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service