Skip to Content
Merck
All Photos(1)

Documents

454454

Sigma-Aldrich

Methoxysuccinyl-Ala-Ala-Pro-Val-p-nitroanilide

≥98% (HPLC), solid, human leukocyte elastase substrate, Calbiochem®

Synonym(s):

Methoxysuccinyl-Ala-Ala-Pro-Val-p-nitroanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H38N6O9
CAS Number:
Molecular Weight:
590.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Methoxysuccinyl-Ala-Ala-Pro-Val-p-nitroanilide, Soluble, specific substrate for human leukocyte elastase.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

ethanol: 1 mg/mL
DMF: 5 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C27H38N6O9/c1-15(2)23(30-18-8-10-19(11-9-18)33(40)41)26(38)31-25(37)20-7-6-14-32(20)27(39)17(4)29-24(36)16(3)28-21(34)12-13-22(35)42-5/h8-11,15-17,20,23,30H,6-7,12-14H2,1-5H3,(H,28,34)(H,29,36)(H,31,37,38)/t16-,17-,20-,23-/m0/s1

InChI key

JWYKDZIEIWGSBN-OSAZLGQLSA-N

General description

Soluble specific substrate for human leukocyte elastase. Is 200-fold more sensitive than the conventional substrate.
Soluble, specific substrate for human leukocyte elastase. Is 200-fold more sensitive than the conventional substrate. Cleavage of substrate can be monitored at ~405 nm.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Substrate for human leukocyte elastase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

MeOSuc-Ala-Ala-Pro-Val-pNA, MeOH

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service