Skip to Content
Merck
All Photos(1)

Key Documents

448106

Sigma-Aldrich

Met Kinase Inhibitor VI, SGX523

The Met Kinase Inhibitor VI, SGX523, also referenced under CAS 1022150-57-7, controls the biological activity of Met Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Met Kinase Inhibitor VI, SGX523, 6-(6-(1-Methyl-1H-pyrazol-4-yl)-[1,2,4]triazolo[4,3-b]pyridazin-3-ylthio)quinoline, SGX523

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13N7S
CAS Number:
Molecular Weight:
359.41
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 2.5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C18H13N7S/c1-24-11-13(10-20-24)16-6-7-17-21-22-18(25(17)23-16)26-14-4-5-15-12(9-14)3-2-8-19-15/h2-11H,1H3

InChI key

BCZUAADEACICHN-UHFFFAOYSA-N

General description

A cell-permeable triazolopyridazine compound that acts as a potent, ATP-competitive (Ki = 2.7 & 23 nM using non-phosphorylated and phosphorylated MET kinase domain, respectively), and highly selective inhibitor against the tyrosine kinase activity of the HGF (hepatocyte growth factor) receptor c-MET (IC50 = 4 nM), while exhibiting little or no activity toward a panel of more than 210 other kinases, including the closely related RTK RON. Shown to inhibit the constitutive (IC50 = 40 nM in GTL16 cells) as well as HGF-stimulated (IC50 = 12 nM in A549 cells) MET autophosphorylation/activation in cultures in vitro and effectively suppress MET-dependent tumor growths in mice in vivo (twice daily oral dosings at 30 mg/kg). X-ray crystallography reveals that SGX523 locks MET in a "DFG-in" conformation with the triazolopyridazine moiety interacting with the hydrophobic side chains of Met1229 and Tyr1248, while Tyr1248 mutations are found to render MET insensitive to SGX523 inhibition both in cell-free and cell-based assays.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months -20°C.

Other Notes

Buchanan, S.G., et al. 2009. Mol. Cancer Ther.8, 3181.
Eder, J.P., et al. 2009. Clin. Cancer Res.15, 2207.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Warren Naselsky et al.
Molecular carcinogenesis, 61(6), 537-548 (2022-03-24)
Transglutaminase 2 (TG2) is an important mesothelioma cancer cell survival protein. However, the mechanism whereby TG2 maintains mesothelioma cell survival is not well understood. We present studies showing that TG2 drives hepatocyte growth factor (HGF)-dependent MET receptor signaling to maintain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service