Skip to Content
Merck
All Photos(1)

Key Documents

445888

Sigma-Aldrich

Metastin (45-54)

A decapeptide with C-terminally amidated LRF-motif that acts as a potent and selective agonist of AXOR12 and hOT7T175 (a novel human G-protein-coupled receptors).

Synonym(s):

Metastin (45-54), KiSS-1 (112-121), Kisspeptin-10, H-YNWNSFGLRF-NH₂

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 134.00

CHF 134.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
CHF 134.00

About This Item

Empirical Formula (Hill Notation):
C63H83N17O14
Molecular Weight:
1302.44
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 134.00


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A decapeptide with C-terminally amidated LRF-motif that acts as a potent and selective agonist of AXOR12 and hOT7T175 (a novel human G-protein-coupled receptors). Induces an increase in intracellular Ca2+ levels in CHO/h175 and CHO/AXOR12:Gqi5 cells and exhibits about 8-fold higher receptor binding affinity (Ki = 42 pM) compared to Metastin (Cat. No. 445885).

Biochem/physiol Actions

Cell permeable: no
Primary Target
AXOR12 and hOT7T175
Product does not compete with ATP.
Reversible: no
Target Ki: 42 pM for AXOR12 and hOT7T175

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Tyr-Asn-Trp-Asn-Ser-Phe-Gly-Leu-Arg-Phe-NH₂

Other Notes

Kotani, M., et al. 2001. J. Biol. Chem.276, 34631.
Ohtaki, T., et al. 2001. Nature411, 613.
Muir, A.I., et al. 2001. J. Biol. Chem.276, 28969.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service