Skip to Content
Merck
All Photos(1)

Key Documents

440345-M

Millipore

Lysozyme, Human Neutrophil

Occurs in specific granules of human polymorphonuclear leukocytes and takes part in antibacterial activity of leukocytes.

Synonym(s):

Lysozyme, Human Neutrophil, N-Acetylmuramyl Hydrolase, Muramidase

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 742.00

CHF 742.00


Estimated to ship on31 May 2025



Select a Size

Change View
1 MG
CHF 742.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

CHF 742.00


Estimated to ship on31 May 2025


description

Merck USA index - 14, 5640

Assay

≥95% (SDS-PAGE)

form

lyophilized

specific activity

≥30,000 Shugar units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

aqueous buffer: soluble
water: soluble

storage temp.

−20°C

SMILES string

N1([C@@H](CCC1)C(=O)N[C@@H](CCC(=O)N)C(=O)NCC(=O)N[C@@H]([C@H](CC)C)C(=O)N[C@@H](CCCN\C(=N\[H])\N)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc4c5c([nH]c4)cccc5)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc2c3c([nH]c2)cccc3)C(=O)N[C@@H](CCCN\C(=N\[H])\N)C(=O)N[C@@H]

InChI

1S/C99H159N37O23/c1-11-50(8)77(132-72(139)46-120-81(145)63(32-33-70(101)137)124-88(152)69-31-21-39-136(69)93(157)68(43-73(140)141)131-84(148)62(29-19-37-116-98(109)110)125-90(154)75(48(4)5)135-91(155)76(49(6)7)133-80(144)57(100)24-16-34-113-95(103)104)92(156)126-60(27-17-35-114-96(105)106)82(146)121-51(9)78(142)129-66(41-54-45-119-59-26-15-13-23-56(54)59)87(151)134-74(47(2)3)89(153)122-52(10)79(143)128-65(40-53-44-118-58-25-14-12-22-55(53)58)85(149)123-61(28-18-36-115-97(107)108)83(147)130-67(42-71(102)138)86(150)127-64(94(158)159)30-20-38-117-99(111)112/h12-15,22-23,25-26,44-45,47-52,57,60-69,74-77,118-119H,11,16-21,24,27-43,46,100H2,1-10H3,(H2,101,137)(H2,102,138)(H,120,145)(H,121,146)(H,122,153)(H,123,149)(H,124,152)(H,125,154)(H,126,156)(H,127,150)(H,128,143)(H,129,142)(H,130,147)(H,131,148)(H,132,139)(H,133,144)(H,134,151)(H,135,155)(H,140,141)(H,158,159)(H4,103,104,113)(H4,105,106,114)(H4,107,108,115)(H4,109,110,116)(H4,111,112,117)/t50-,51-,52-,57-,60-,61-,62-,63-,64-,65-,66-,67-,68-,69-,74-,75-,76-,77-/m0/s1

InChI key

ZJCXKXFAOCLSRV-RYLVIXIQSA-N

General description

Native lysozyme from human neutrophils. Occurs in specific granules of human polymorphonuclear leukocytes and takes part in antibacterial activity of leukocytes. Hydrolyzes β1,4 linkages between N-acetylmuramic acid (2-acetamido-2-deoxy-3-O-[1′-carboxymethyl]-glucopyranose) residues in chitin.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One Shugar unit is defined as the amount of enzyme that will cause a decrease in absorbance of 0.001 at 450 nm per min due to lysis of Micrococcus lysodeikticus at 37°C, pH 7.0.

Physical form

Lyophilized from 100 mM NaCl, 50 mM sodium acetate buffer, pH 6.0.

Preparation Note

Prepared from blood that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 3 days at 4°C or for up to 1 year at -20°C.

Other Notes

Lollike, K., et al. 1995. Leukemia 9, 206.
Gordon, L.I., et al. 1979. J. Clin. Invest. 64, 226.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service