Skip to Content
Merck
All Photos(1)

Key Documents

437600

Millipore

LPD-12

A water soluble, amphiphilic α-helical lipopeptide detergent (LPD) that forms small micelles and effectively mimics lipid bilayers.

Synonym(s):

LPD-12, Lipopeptide Detergent-12, Ac-AO(C₁₂)AEAAEKAAKYAAEAAEKAAKAO(C₁₂)A-NH₂

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C131H224N32O37
Molecular Weight:
2839.37
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

CMC

<1 μM

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A water soluble, amphiphilic α-helical lipopeptide detergent (LPD) that forms small micelles and effectively mimics lipid bilayers (solubility in PBS ~ 40 mM; micelle MW ~ 23,100; aggregation number ~ 8.1; partial specific volume ~ 0.77 cm3/g). Shown to not only solubilize but also stabilize bacteriorhodopsin, lactose permease and E. coli PagP protein ([LPD] ~ 250 µM and [protein] ~ 22 µM) by forming complexes and precluding aggregation. Useful for NMR and X-ray structural studies of membrane proteins.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Sequence

Ac-Ala-Orn(dodecanoyl)-Ala-Glu-Ala-Ala-Glu-Lys-Ala-Ala-Lys-Tyr-Ala-Ala-Glu-Ala-Ala-Glu-Lys-Ala-Ala-Lys-Ala-Orn(dodecanoyl)-Ala-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

McGregor, C.L., et al. 2003. Nat. Biotechnol.21, 171.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service