Skip to Content
Merck
All Photos(2)

Key Documents

412512

Sigma-Aldrich

IRE1 Inhibitor III, 4μ8C

IRE1 Inhibitor III, CAS 14003-96-4, is a cell-permeable. Covalent inhibitor of IRE1 RNase activity (IC₅₀ = 550 and 45 nM, respectively, with 0 & 16 min preincubation in RNA cleavage assays).

Synonym(s):

IRE1 Inhibitor III, 4μ8C, 8-Formyl-7-hydroxy-4-methylcoumarin, 7-Hydroxy-4-methyl-2-oxo-2H-chromene-8-carbaldehyde, ER-to-Nucleus Signaling 1 Inhibitor III, Inositol-Reguiring Protein 1 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8O4
CAS Number:
Molecular Weight:
204.18
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(C=C1C)OC2=C1C=CC(O)=C2C=O

InChI

1S/C11H8O4/c1-6-4-10(14)15-11-7(6)2-3-9(13)8(11)5-12/h2-5,13H,1H3

InChI key

RTHHSXOVIJWFQP-UHFFFAOYSA-N

General description

A cell-permeable coumarin o-hydroxyaldehyde compound that inhibits IRE1 RNase activity in a time- and dose-dependent manner (IC50 = 550, 230, 180, 100, and 45 nM, respectively, with 0, 2, 4, 8, 16, min drug preincubation in FRET-based RNA cleavage assays) by covalently targeting IRE1 Lys907 via Schiff base formation, effectively preventing ER stress-induced site-specific mRNA splicing as well as RIDD (Regulated IRE1-Dependent Degradation) mRNA degradation (IC50 = 6.9 and 4.1 µM, respectively, against Xbp1 splicing and Scara3 degradation) in MEF cultures following Tunicamycin (Cat. No. 654380) treatment. Also demonstrated to inhibit ER capacity expansion (Effective conc. 32 µM) and amylase secretion (IC50<2 µM) upon stress induction by Dexamethasone (Cat. No. 265005) treatment in rat AR42J tumoral acinar pancreatic cells. Structural analysis reveals that the reduced water accessibility to Lys907 in IRE1 native conformation accounts for the unusual stability of Lys907 Schiff base formation and forms the basis of selective IRE1 RNase inhibition by 4μ8C and STF083010 (Cat. No. 412510). Although 4μ8C, but not STF083010, is also shown to inhibit IRE1 autophosphorylation by Schiff base formation with IRE1 Lys599 in the absence of ADP, cellular nucleotide prevents 4μ8C from targeting IRE1 Lys599 and inhibiting IRE1 kinase activity intracellularly.
A cell-permeable coumarin o-hydroxyaldehyde that inhibits IRE1 RNase activity in a time- and dose-dependent manner (IC50 = 550 and 45 nM, respectively, with 0 and 16 min drug preincubation in RNA cleavage assays) by covalently targeting IRE1 Lys907 via Schiff base formation, effectively preventing ER stress-induced site-specific mRNA splicing as well as RIDD (Regulated IRE1-Dependent Degradation) mRNA degradation (IC50 = 6.9 and 4.1 µM, respectively, against Xbp1 splicing and Scara3 degradation) in MEF cultures following Tunicamycin (Cat. No. 654380) treatment. Comparing to STF083010 (Cat. No. 412510), 4μ8C is also shown to inhibit IRE1 autophosphorylation in cell-free assays via Schiff base formation with IRE1 Lys599 in the absence of ADP, however cellular nucleotide prevents 4μ8C from targeting IRE1 Lys599 intracellularly.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IRE1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months.

Other Notes

Cross, B.C.S., et al. 2012. Proc. Natl. acad. Sci. USA109, E869.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HIF1I?-dependent hypoxia response in myeloid cells requires IRE1I?.
Mawambo, et al.
Journal of Neuroinflammation, 20, 145-145 (2023)
Aparajita Madhavan et al.
Nature communications, 13(1), 608-608 (2022-02-03)
In obesity, signaling through the IRE1 arm of the unfolded protein response exerts both protective and harmful effects. Overexpression of the IRE1-regulated transcription factor XBP1s in liver or fat protects against obesity-linked metabolic deterioration. However, hyperactivation of IRE1 engages regulated IRE1-dependent
Methionine consumption by cancer cells drives a progressive upregulation of PD-1 expression in CD4 T cells.
Pandit, et al.
Nature Communications, 14, 2593-2593 (2023)
Mariana Marques et al.
iScience, 27(3), 109100-109100 (2024-02-26)
Influenza A virus (IAV) employs multiple strategies to manipulate cellular mechanisms and support proper virion formation and propagation. In this study, we performed a detailed analysis of the interplay between IAV and the host cells' proteostasis throughout the entire infectious
José P Guirao-Abad et al.
mSphere, 5(5) (2020-10-23)
The unfolded protein response (UPR) is a signaling network that maintains homeostasis of the endoplasmic reticulum (ER). In the human-pathogenic fungus Aspergillus fumigatus, the UPR is initiated by activation of an endoribonuclease (RNase) domain in the ER transmembrane stress sensor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service