Skip to Content
Merck
All Photos(1)

Key Documents

411423

Sigma-Aldrich

Mouse Anti-Human IgA, Secretory (HP6141)

liquid, clone HP6141, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

400 μG
CHF 514.00

CHF 514.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
400 μG
CHF 514.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

CHF 514.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

HP6141, monoclonal

form

liquid

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

General description

Purified mouse monoclonal IgG. Recognizes human IgA secretory subunit.
This Mouse Anti-Human IgA, Secretory (HP6141) is validated for use in ELISA for the detection of Human IgA, Secretory.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Can be used as a capture antibody for ELISA at 10 µg/ml in PBS without carrier protein. Antibody should be titrated for optimal results in individual systems.



Cross-reactivity by ELISA against human myeloma proteins:

Human IgG: <0.01%

Human IgE: <0.01%

Human IgM: <0.01%

Human IgA1: <0.01%

Human IgA2: <0.01%

Human secretory IgA: 100%
Hamilton, R.G., 1990. Ann. Biol. Clin.48, 473.
Fasullo, F.J., et al. 1989. Clin. Chem.35, 364.
Reimer, C.B., et al. 1989. Immunol. Letters21, 209.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alba Escalera et al.
iScience, 27(3), 109210-109210 (2024-03-04)
Despite multiple research efforts to characterize coronavirus disease 2019 (COVID-19) in humans, there is no clear data on the specific role of mucosal immunity on COVID-19 disease. Here, we longitudinally profile the antibody response against severe acute respiratory syndrome coronavirus
Hailey Hornsby et al.
Nature communications, 14(1), 5065-5065 (2023-08-22)
Pronounced immune escape by the SARS-CoV-2 Omicron variant has resulted in many individuals possessing hybrid immunity, generated through a combination of vaccination and infection. Concerns have been raised that omicron breakthrough infections in triple-vaccinated individuals result in poor induction of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service