Skip to Content
Merck
All Photos(1)

Key Documents

400053

Millipore

Hygromycin B, Streptomyces sp., Sterile-Filtered Solution in 25 mM HEPES, Cell Culture-Tested

Hygromycin B is an aminoglycoside antibiotic that inhibits the growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis by interfering with the translocation of the 70S ribosome and causes misreading of mRNA.

Synonym(s):

Hygromycin B, Streptomyces sp., Sterile-Filtered Solution in 25 mM HEPES, Cell Culture-Tested

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H37N3O13
CAS Number:
Molecular Weight:
527.52
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥90% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C20H37N3O13/c1-23-7-2-5(21)9(26)15(10(7)27)33-19-17-16(11(28)8(4-25)32-19)35-20(36-17)18(31)13(30)12(29)14(34-20)6(22)3-24/h5-19,23-31H,2-4,21-22H2,1H3/t5-,6+,7+,8-,9+,10-,11+,12-,13-,14-,15-,16+,17+,18-,19+,20?/m1/s1

InChI key

GRRNUXAQVGOGFE-KPBUCVLVSA-N

General description

Hygromycin B is an aminoglycoside antibiotic, inhibits the growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis by interfering with the translocation of the 70S ribosome and causes misreading of mRNA. Resistance to hygromycin B is conferred by a gene whose product, hygromycin B phospho-transferase that phosphorylates and inactivates the drug. Hygromycin B has been used to select mutants carrying the hygromycin B resistance gene in a wide variety of cells including bacterial, protozoal, yeast, fungal, plant, and mammalian cells. Typical concentrations used for mammalian cell selection are between 0.05 to 1 mg/ml. The appropriate concentration for use must be determined empirically for each cell type.

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Physical form

50 mg/ml active antibiotic in sterile-filtered 25 mM HEPES, pH 7.2.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Sugimoto, K., et al. 1994. Plant J.5, 863.
Lee, M.G., and van der Ploeg, L.H. 1991. Gene105, 255.
Salauze, D., et al. 1990. Antimicrob. Agents Chemother.34, 1915.
Cullen, D., et al. 1987. Gene57, 21.
Santerre, R.F., et al. 1984. Gene30, 147.
Rao, S.N., et al. 1983. Antimicrob. Agents Chemother.24, 689.
Gonzalez, A., et al. 1978. Biochim. Biophys. Acta521, 459.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jong-Ho Lee et al.
Frontiers in oncology, 10, 211-211 (2020-03-21)
Metabolism plays a critical role in direct regulation of a variety of cellular activities via metabolic enzymes and metabolites. Here, we demonstrate that phosphofructokinase 1 platelet isoform (PFKP), which catalyzes a rate-limiting reaction in glycolysis, promotes EGFR activation-induced nuclear translocation

Articles

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service