Skip to Content
Merck
All Photos(1)

Key Documents

382159-M

Millipore

PhosphoDetect Anti-Histone H3 (pSer¹⁰) (7-20) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

Xenopus, Tetrahymena sp., human, Drosophila, Caenorhabditis elegans

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer10)

Gene Information

human ... H3C1(8350)

General description

Protein A purified rabbit polyclonal antibody. Recognizes the ~17 kDa histone H3 protein phosphorylated at Ser10.
Recognizes the ~17 kDa histone H3 protein phosphorylated at Ser10.
This PhosphoDetect Anti-Histone H3 (pSer¹⁰) (7-20) Rabbit pAb is validated for use in Immunoblotting, ICC, IHC, IP for the detection of Histone H3 (pSer¹⁰) (7-20).

Immunogen

Human
a synthetic phosphopeptide [ARKpSTGGKAPRKQL(C)] corresponding to amino acids 7-20 surrounding the Ser¹⁰ phosphorylation site of human histone H3, conjugated to KLH

Application

Immunoblotting (0.5 µg/ml)

Immunocytochemistry (5 µg/ml)

Immunohistochemistry (see comments)

Immunoprecipitation (4 µg/10 µg acid-extracted proteins from colcemid-arrested HeLa cells)

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 100 mM Tris-glycine, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Acid-extracted proteins from colcemid-arrested HeLa cells

Other Notes

Ajiro, K., et al. 1996. J. Biol. Chem.271, 13197.
Chadee, D.N., et al. 1995. J. Biol. Chem.270, 20098.
Mahadevan, L.C., et al. 1991. Cell65, 775.
For immunocytochemistry, colcemid-arrested HeLa cells were fixed with 1% paraformaldehyde. Also detects histone H3 in Tetrahymena micronuclei. The antibody has been reported to detect mitotic cells in Xenopus, Caenorhabditis elegans, and Drosophila embryos. Does not cross-react with unphosphorylated histone H3. This antibody has also been reported to work for immunohistochemistry. Variables associated with assay conditions will dictate the optimal working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service