Skip to Content
Merck
All Photos(1)

Documents

377980

Sigma-Aldrich

Hispidin

A potent, cell permeable, and reversible inhibitor of protein kinase Cβ isoform (PKCβ; IC₅₀ = 2 µM).

Synonym(s):

Hispidin, 6-(3,4-Dihydroxystyrl)-4-hydroxy-2-pyrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O5
CAS Number:
Molecular Weight:
246.22
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥90% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C13H10O5/c14-9-6-10(18-13(17)7-9)3-1-8-2-4-11(15)12(16)5-8/h1-7,15-17H/b3-1+

InChI key

QISBNGLMCHHHFG-HNQUOIGGSA-N

General description

A potent, cell permeable, and reversible inhibitor of protein kinase Cβ isoform (PKCβ; IC50 = 2 µM). Has stronger cytotoxic effects on cancer cells than on normal cells in vitro.
A potent, cell permeable, and reversible inhibitor of protein kinase Cβ isoform (PKCβ; IC50 = 2 µM). Has stronger cytotoxic effects on cancer cells than on normal cells in vitro.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKCβ
Product does not compete with ATP.
Reversible: yes
Target IC50: 2 µM against PKCβ

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gonindard, C., et al. 1997. Cell Biol. Toxicol. 13, 141.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service