Skip to Content
Merck
All Photos(1)

Key Documents

3750-OP

Sigma-Aldrich

DNase I

OmniPur® Grade, from Bovine Pancreas

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Quality Level

form

crystalline powder

manufacturer/tradename

Calbiochem®

color

white to off-white

shipped in

ambient

General description

Note: 1PC = 50,000 Units

Enzyme for hydrolysis of DNA for critical RNA isolation applications. Reconstitute the enzyme to a concentration of at least 2 mg/ml in a 50% Glycerol solution (Cat. No. 4750) with 20 mM Tris-HCl, pH 7.5 (Cat. No. 9285) and 1 mM MgCl2 (Cat. No. 5985). The solution may be stored at -20°C for at least 1 year.

Specifications:
Activity (Kunitz, 25°C): ≥1800U/mg
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service