Skip to Content
Merck
All Photos(1)

Key Documents

374834

Sigma-Aldrich

Hemoglobin, Bovine Erythrocytes

Hemoglobin, Bovine Erythrocytes, CAS 9008-02-0, is an iron-containing oxygen-transport metalloprotein. Suitable as a substrate for proteases, such as papain, pepsin, and trypsin.

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
CHF 119.00

CHF 119.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
10 G
CHF 119.00

About This Item

UNSPSC Code:
12352202

CHF 119.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 4648

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Native hemoglobin from bovine erythrocytes. Excellent substrate for proteases, such as papain, pepsin, and trypsin.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Antonini, E., et al. 1981. Methods Enzymol. 76, 1.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service