Skip to Content
Merck
All Photos(1)

Key Documents

374817

Sigma-Aldrich

Hemocyanin, Keyhole Limpet, Megathura crenulata, Glycerol

Hemocyanin, Keyhole Limpet, Megathura crenulata, Glycerol, is a large, multi-subunit, oxygen-carrying, metalloprotein that is used as a carrier protein in the production of antibodies.

Synonym(s):

Hemocyanin, Keyhole Limpet, Megathura crenulata, Glycerol, KLH

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 164.00
250 MG
CHF 426.00
1 G
CHF 1’660.00

CHF 164.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
CHF 164.00
250 MG
CHF 426.00
1 G
CHF 1’660.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

CHF 164.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 4647

Assay

>90% (size exclusion chromatography)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear blue-gray

shipped in

ambient

storage temp.

2-8°C

General description

Carrier protein that has many available primary amines to facilitate protein conjugation. Used to immunize animals to elicit antibodies to small molecules (haptens) by covalent conjugation. Protein: ≥10 mg/ml. Sold on the basis of protein content.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM MgSO₄, 5 mM BES, 50% glycerol, 0.9% sodium chloride, pH 6.5.

Preparation Note

Further dilute with PBS or physiological saline. Material can be dialyzed to remove the glycerol or used as is for conjugation with glutaraldehyde, carbodiimide, or mixed anhydrides.KLH is an extremely large protein complex that has a tendency to aggregate and precipitate from solutions. Use a buffer when diluting KLH and maintain the magnesium concentration at ≥10 mM. Store KLH solutions at 4°C. DO NOT freeze, vortex or agitate to dissolve KLH. Freezing or agitating KLH will cause protein aggregation and precipitation.

Other Notes

Hayashi, K., et al. 1992. Virchows Archiv. B Cell Pathol.63, 37.
Shuler, K.R., et al. 1992. J. Immunol. Methods156, 137.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service