Skip to Content
Merck
All Photos(2)

Key Documents

373277

Sigma-Aldrich

Hh Signaling Antagonist XI, MRT-14

The Hh Signaling Antagonist XI, MRT-14 controls the biological activity of Hh pathway. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

Hh Signaling Antagonist XI, MRT-14

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23N3O6
CAS Number:
Molecular Weight:
449.46
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NC(NC1=CC(NC(C2=CC=CC=C2)=O)=CC=C1)=O)C3=CC(OC)=C(C(OC)=C3)OC

General description

A cell-permeable acylurea compound that acts as an antagonistic Smo ligand (IC50 = 120 nM against 5 nM Bodipy-cyclopamine binding to murine Smo-transfected HEK293 cells) and is more effective than Cyclopamine (Cat. No. 239803) in inhibiting ShhN-induced Gli transcription in Shh-light 2 cell assays (IC50 = 0.16 and 0.3 µM, respectively) and in blocking 0.1 µM SAG- (Cat. Nos. 566660 & 566661) stimulated differentiation of C3H10T1/2 murine mesenchymal stem cells into osteoblasts (IC50 = 0.13 and 0.62 µM, respectively).
The Hh Signaling Antagonist XI, MRT-14 controls the biological activity of Hh pathway. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Manetti, F., et al. 2010. Mol. Pharmacol.78, 658.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service