Skip to Content
Merck
All Photos(1)

Key Documents

371751

Sigma-Aldrich

Anti-Gqα- and G₁₁α-Subunits, C-Terminal Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
CHF 401.00

CHF 401.00


Estimated to ship on19 April 2025


Request a Bulk Order

Select a Size

Change View
50 μL
CHF 401.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 401.00


Estimated to ship on19 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... GNAI1(2770)

General description

Anti-Gqα- and G₁₁α-Subunits, C-Terminal, rabbit polyclonal, recognizes the Gqα- and G₁₁α-subunits. Does not cross-react with other G-proteins. Validated for use in ELISA, WB, and IP.
Protein A affinity purified rabbit polyclonal antibody. Recognizes Gqα and G11α subunit proteins.
Recognizes the Gqα-and G11α-subunits. Does not cross-react with other G-proteins of the non-pertussis toxin-sensitive family.

Immunogen

a synthetic peptide (QLNLKEYNLV) corresponding to amino acids at the C-terminus of mammalian Gqα and G₁₁α, conjugated to KLH

Application

ELISA (see comments)

Immunoblotting (1:1000)

Immunoprecipitation (see comments)

Warning

Toxicity: Standard Handling (A)

Other Notes

The decapeptide corresponds to a defined C-terminal sequence found in both Gqα and G11α. Does not cross-react with other G proteins of the non-pertussis toxin sensitive family. This antibody has also been reported to work for ELISA and immunoprecipitation. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Hansen et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 23(28), 9328-9339 (2003-10-17)
The olfactory epithelium of fish contains three intermingled types of olfactory receptor neurons (ORNs): ciliated, microvillous, and crypt. The present experiments were undertaken to test whether the different types of ORNs respond to different classes of odorants via different families
Heather D Bracken-Grissom et al.
Scientific reports, 10(1), 4485-4485 (2020-03-13)
Extraocular photoreception, the ability to detect and respond to light outside of the eye, has not been previously described in deep-sea invertebrates. Here, we investigate photosensitivity in the bioluminescent light organs (photophores) of deep-sea shrimp, an autogenic system in which
Merry C Clark et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 143(3), 294-301 (2006-01-24)
Dopamine (DA) differentially modulates identified neurons in the crustacean stomatogastric nervous system (STNS). While the electrophysiological actions of DA have been well characterized, little is known about the dopaminergic transduction cascades operating in this system. As a first step toward

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service