Skip to Content
Merck
All Photos(1)

Documents

370695

Sigma-Aldrich

GW1929

A potent, tyrosine-based peroxisome proliferator-activated receptor γ (PPARγ) agonist (EC₅₀ = 13 nM for murine receptor and 6.2 nM for human receptor in cell-based transactivation assays).

Synonym(s):

GW1929, PPAR Agonist V, PPARγ Agonist IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H29N3O4
CAS Number:
Molecular Weight:
495.57
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C30H29N3O4/c1-33(28-13-7-8-18-31-28)19-20-37-24-16-14-22(15-17-24)21-27(30(35)36)32-26-12-6-5-11-25(26)29(34)23-9-3-2-4-10-23/h2-18,27,32H,19-21H2,1H3,(H,35,36)/t27-/m0/s1

InChI key

QTQMRBZOBKYXCG-MHZLTWQESA-N

General description

A potent, tyrosine-based peroxisome proliferator-activated receptor γ (PPARγ) agonist (EC50 = 13 nM for murine PPARγ and 6.2 nM for human PPARγ in cell-based transactivation assays).
A potent, tyrosine-based peroxisome proliferator-activated receptor γ (PPARγ) agonist (EC50 = 13 nM for murine receptor and 6.2 nM for human receptor in cell-based transactivation assays).

Biochem/physiol Actions

Cell permeable: no
EC50 = 13 nM as peroxisome proliferator-activated receptor γ (PPARγ) agonist for murine receptor and 6.2 nM for human receptor in cell-based transactivation assays
Primary Target
Peroxisome proliferator-activated receptor γ (PPARγ) agonist
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Han, S., et al. 2004. Clin. Cancer Res.10, 1911.
Willson, T.M., et al. 2000. J. Med. Chem.43, 527.
Brown, K.K., et al. 1999. Diabetes48, 1415.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service