Skip to Content
Merck
All Photos(1)

Key Documents

368055

Sigma-Aldrich

Granzyme B Inhibitor II

The Granzyme B Inhibitor II, also referenced under CAS 1258003-96-1, controls the biological activity of Granzyme B. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Granzyme B Inhibitor II, Caspase-8 Inhibitor I, Ac-IETD-CHO

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 119.00

CHF 119.00


Estimated to ship on29 May 2025



Select a Size

Change View
1 MG
CHF 119.00

About This Item

Empirical Formula (Hill Notation):
C21H34N4O10
CAS Number:
Molecular Weight:
502.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 119.00


Estimated to ship on29 May 2025


Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N([C@@H]([C@@H](O)C)C(=O)N[C@@H](CC(=O)O)C=O)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)C)[C@H](CC)C)CCC(=O)O

InChI key

AXTKTZHLZLOIIO-WBOIFEBMSA-N

General description

A potent, reversible inhibitor of granzyme B and caspase-8 (Ki = 1 nM). Also inhibits caspase-1 (<6 nM), caspase-6 (5.6 nM), and caspase-10 (27 nM).
Potent, reversible granzyme B and caspase-8 inhibitor.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: yes
Target Ki: 1 nM against caspase-8

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Ile-Glu-Thr-Asp-CHO

Other Notes

Gao, C.F., et al. 2001. Exp. Cell. Res.265, 145.
Garcia-Calvo, M., et al. 1998. J. Biol. Chem.273, 32608.
Han, Z., et al. 1997. J. Biol. Chem. 272, 13422.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdulla Berjis et al.
Nature communications, 15(1), 3937-3937 (2024-05-11)
Human natural killer (NK) cell-based therapies are under assessment for treating various cancers, but cryopreservation reduces both the recovery and function of NK cells, thereby limiting their therapeutic feasibility. Using cryopreservation protocols optimized for T cells, here we find that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service