Skip to Content
Merck
All Photos(1)

Key Documents

345996

Sigma-Aldrich

Glial Fibrillary Acidic Protein, Human Brain

Synonym(s):

GFAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥90% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

distilled water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Native, glial fibrillary acidic protein purified from human brain. Marker of sclerotic plaques in brain and gliosed brain. Isolated from normal human brain. A component of astroglial intermediate filaments. Forms glial filaments in astrocytes in the central nervous system and in some Schwann cells in peripheral nerves.



Molecular Weight: 43,000-49,000

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 16 mM Sodium Bicarbonate, pH 8.5.

Preparation Note

Prepared from tissues of individuals that have been shown by certified tests to be negative for HbsAg, HIV-1 and HIV-2 antibodies, HCV, and syphilis.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Anderiva, M., et al. 2001. Glia35, 189.
Masood, K., et al. 1993. J. Neurochem. 61, 160.
Liem, R.K., and Hutchison, S.B. 1982. Biochemistry21, 3221.
Dahl, D. 1973. Brain Res.57, 343.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald J Ellis et al.
Journal of neurology and psychology, 9(1) (2022-01-01)
Astrocytes become activated with certain infections, and this might alter the brain to trigger or worsen depressed mood. Indeed, astrocytes are chronically activated in people with HIV infection (PWH), who are much more frequently depressed than people without HIV (PWoH).
John E Buonora et al.
Frontiers in neurology, 6, 68-68 (2015-04-15)
Important challenges for the diagnosis and monitoring of mild traumatic brain injury (mTBI) include the development of plasma biomarkers for assessing neurologic injury, monitoring pathogenesis, and predicting vulnerability for the development of untoward neurologic outcomes. While several biomarker proteins have
Mohamed B Abou-Donia et al.
Brain sciences, 10(9) (2020-09-10)
For the past 30 years, there has been a lack of objective tools for diagnosing Gulf War Illness (GWI), which is largely characterized by central nervous system (CNS) symptoms emerging from 1991 Gulf War (GW) veterans. In a recent preliminary
Mohamed B Abou-Donia et al.
Brain sciences, 11(2) (2021-01-28)
Veterans from the 1991 Gulf War (GW) have suffered from Gulf War illness (GWI) for nearly 30 years. This illness encompasses multiple body systems, including the central nervous system (CNS). Diagnosis and treatment of GWI is difficult because there has
Sharmilee Gnanapavan et al.
PloS one, 8(8), e70019-e70019 (2013-08-13)
Lamotrigine trial in SPMS was a randomised control trial to assess whether partial blockade of sodium channels has a neuroprotective effect. The current study was an additional study to investigate the value of neurofilament (NfH) and other biomarkers in predicting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service