Skip to Content
Merck
All Photos(1)

Key Documents

345759

Sigma-Aldrich

Anti-Ganglioside GM₂ Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 647.00

CHF 647.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
CHF 647.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 647.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity (predicted by homology)

all

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Ganglioside GM₂, rabbit polyclonal recognizes ganglioside GM₂ in a wide variety of species. It is validated for use in ELISA and TLC immunoblotting.
Rabbit polyclonal antibody supplied as undiluted serum. Recognizes the GM2 ganglioside.
Recognizes ganglioside GM2.

Immunogen

purified, human ganglioside GM₂

Application

ELISA (1:200)

TLC Immunoblotting (1:100, colorimetric)

Warning

Toxicity: Standard Handling (A)

Other Notes

Does not cross-react with other carbohydrate epitopes. Variables associated with assay conditions will dictate the optimal working dilution.
Yoshino, H., et al. 1993. J. Neurochem. 61, 658.
Kusunoki, S., et al. 1987. Neurology 37, 1795.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuehong Chen et al.
Journal of molecular medicine (Berlin, Germany), 96(12), 1359-1373 (2018-10-21)
Tay-Sachs disease (TSD) is a lethal lysosomal storage disease (LSD) caused by mutations in the HexA gene, which can lead to deficiency of β-hexosaminidase A (HexA) activity and consequent accumulation of its substrate, GM2 ganglioside. Recent reports that progranulin (PGRN)
Aleksandra Somogyi et al.
International journal of molecular sciences, 19(2) (2018-02-23)
Juvenile neuronal ceroid lipofuscinosis (JNCL) is caused by mutations in the CLN3 gene. Most JNCL patients exhibit a 1.02 kb genomic deletion removing exons 7 and 8 of this gene, which results in a truncated CLN3 protein carrying an aberrant
Erica Tagliatti et al.
Immunity, 57(1), 86-105 (2024-01-02)
Triggering receptor expressed on myeloid cells 2 (Trem2) is a myeloid cell-specific gene expressed in brain microglia, with variants that are associated with neurodegenerative diseases, including Alzheimer's disease. Trem2 is essential for microglia-mediated synaptic refinement, but whether Trem2 contributes to
Meredith K Loth et al.
Neurobiology of disease, 85, 174-186 (2015-11-08)
Translocator protein (18 kDa), formerly known as the peripheral benzodiazepine receptor (PBR), has been extensively used as a biomarker of active brain disease and neuroinflammation. TSPO expression increases dramatically in glial cells, particularly in microglia and astrocytes, as a result

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service