Skip to Content
Merck
All Photos(1)

Key Documents

344050

Sigma-Aldrich

Anti-Fodrin Mouse mAb (Fod009)

liquid, clone Fod009, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
CHF 473.00

CHF 473.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
CHF 473.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 473.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

Fod009, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat, human, bovine, canine, porcine, rabbit, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... SPTAN1(6709)

General description

Mouse monoclonal antibody purified by ammonium sulfate precipitation and DEAE column chromatography. Recognizes the ~240 kDa α-subunit of intracellular and plasma membrane-associated fodrin.
Recognizes the ~240 kDa intracellular and plasma membrane associated β subunit of fodrin.
This Anti-Fodrin Mouse mAb (Fod009) is validated for use in ELISA, Flow cytometry, Immunoblotting, Immunoprecipitation for the detection of Fodrin.

Immunogen

Bovine
purified bovine brain fodrin

Application

ELISA (1:1000)

Flow cytometry (1:500)

Immunoblotting (1:200)

Immunohistochemistry (1:100)

Immunoprecipitation (1:100)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 10 mM sodium phosphate, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Lokeshwar, V.B., and Bourguignon, L.Y.W. 1992. J. Biol. Chem. 267, 22073.
Bourguignon, L.Y., and Bourguignon, G.J. 1984. Int. Rev. Cytol.87, 195.
Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Libin Liu et al.
Molecular biology of the cell, 17(10), 4249-4256 (2006-07-28)
Fodrin or nonerythroid spectrin is an abundant component of the cortical cytoskeletal network in rat adipocytes. Fodrin has a highly punctate distribution in resting cells, and insulin causes a dramatic remodeling of fodrin to a more diffuse pattern. Insulin-mediated remodeling
Ke Jin et al.
Cancer prevention research (Philadelphia, Pa.), 9(4), 324-334 (2016-02-07)
Aristolochic acid I (AAI) existing in plant drugs from Aristolochia species is an environmental human carcinogen associated with urothelial cancer. Although gene association network analysis demonstrated gene expression profile changes in the liver of human TP53 knock-in mice after acute

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service