Skip to Content
Merck
All Photos(1)

Key Documents

341599

Sigma-Aldrich

FGF-10, Human, Recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥97% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, human fibroblast growth factor 10 (FGF-10) consisting of amino acids 137-208 and expressed in E. Coli. Mature human FGF-10, a ~20 kDa protein, contains a serine-rich region near its N-terminus sharing 93% amino acid sequence identity with mouse FGF-10 and 96% with rat FGF-10.

Biochem/physiol Actions

ED₅₀ = 20-100 ng/ml measured in a cell proliferation assay using 4MBr-5 monkey epithelial cells.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm sterile-filtered MOPS, Na₂SO₄, EDTA, DTT, BSA.

Reconstitution

Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 100 µg/ml in sterile PBS containing at least 0.1% human or bovine serum albumin.

Other Notes

Beenken, A. and M. Mohammadi. 2009. Nat. Rev. Drug Discov.8, 235.
Nomura, S., et al. 2008. Br. J. Cancer99, 305.
Zhang, X., et al. 2006. J. Biol. Chem.281, 15694.
Beer, H.D., et al. 2005. Oncogene24, 5269.
Jaskoll, T., et al. 2005. BMC Dev. Biol.5, 11.
Rice, R., et al. 2004. J. Clin. Invest.113, 1692.
Donjacour, A.A., et al. 2003. Dev. Biol.261, 39.
Mailleux, A.A., et al. 2002. Development129, 53.
Sakaue, H., et al. 2002. Genes Dev.16, 908.
Makarenkova, H.P., et al. 2000. Development127, 2563.
Makarenkova, H.P., et al. 2000. Development127, 2563.
Pirvola, U., et al. 2000. J. Neurosci.20, 6125.
Weaver, M., et al. 2000. Development127, 2695.
Igarashi, M., et al. 1998. J. Biol. Chem.273, 13230.
Min, H., et al. 1998. Genes Dev.12, 3156.
Bellusci, S., et al. 1997. Development124, 4867.
Emoto, H., et al. 1997. J. Biol. Chem.272, 23191.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service