Skip to Content
Merck
All Photos(2)

Key Documents

324897

Sigma-Aldrich

eIF-2α Inhibitor, Salubrinal

InSolution, ≥95%

Synonym(s):

InSolution eIF-2α Inhibitor, Salubrinal, N-(2,2,2-Tichloro-1-(3-quinolin-8-ylthioureido)ethyl)cinnamamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17Cl3N4OS
CAS Number:
Molecular Weight:
479.81
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

dry ice

storage temp.

−70°C

SMILES string

S=C(NC(NC(/C=C/C1=CC=CC=C1)=O)C(Cl)(Cl)Cl)NC2=CC=CC3=C2N=CC=C3

General description

A cell-permeable and selective inhibitor of the phosphatase complexes that dephosphorylate eukaryotic translation initiation factor 2 subunit α (eIF-2α). Shown to offer protection against ER stress-induced apoptosis (EC50 ~15 µM in PC12 cells stimulated with 750 ng/ml of Tunicamycin (Cat. No. 654380). Prevents HSV replication both in Vero cells (IC50 ~3 µM for reducing plaque formation) in vitro and in a mouse cornea infection model in vivo. The solid form of this compound (Cat. No. 324895) is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
eIF-2α

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 50 mM (5 mg/208 µL) solution of eIF-2α Inhibitor, Salubrinal (Cat. No. 324895) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -70°C.

Other Notes

Boyce, M., et al. 2005. Science307, 935.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (refers to pure substance)

Flash Point(C)

87 °C - closed cup - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Da Shi et al.
PLoS pathogens, 19(3), e1011201-e1011201 (2023-03-09)
Autophagy plays an important role in the infectious processes of diverse pathogens. For instance, cellular autophagy could be harnessed by viruses to facilitate replication. However, it is still uncertain about the interplay of autophagy and swine acute diarrhea syndrome coronavirus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service