Skip to Content
Merck
All Photos(1)

Key Documents

324699

Sigma-Aldrich

Elastase Substrate IV, Colorimetric

A specific substrate for pancreatic elastase.

Synonym(s):

Elastase Substrate IV, Colorimetric

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H34N6O9
Molecular Weight:
562.57
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated

color

white to off-white

solubility

ethanol: 1 mg/mL
water: soluble

shipped in

ambient

storage temp.

−20°C

General description

A specific substrate for pancreatic elastase (Km = 100 µM; Kcat/Km = 35,000 s-1 M-1 for rat pancreatic elastase; Km = 30 µM; Kcat/Km = 351,000 s-1 M-1 for porcine pancreatic elastase). Cleavage of substrate can be monitored at ~405 nm.



Suggested protocol: see Largman, C. (1983)
A specific substrate for pancreatic elastase (km = 100 µM; kcat/ Km = 35,000 M-1 s-1 for rat pancreatic elastase, Km = 30 µM; kcat/Km = 35,000 M-1 s-1 for porcine). Cleavage of substrate can be monitored at ~405 nm.

Biochem/physiol Actions

Cell permeable: no
Primary Target
A specific substrate for pancreatic elastase
Product does not compete with ATP.
Reversible: no
km = 100 µM; kcat/ Km = 35,000 M-1 s-1 for rat pancreatic elastase; Km = 30 µM; kcat/Km = 35,000 M-1 s-1 for porcine

Warning

Toxicity: Standard Handling (A)

Sequence

Suc-Ala-Ala-Pro-Abu-pNA (Abu = L-α-Aminobutyric Acid)

Physical form

45 mg D-mannitol and 5 mg substrate. Sold on the basis of substrate content.

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 3 months at 4°C.

Other Notes

Largman, L. 1983. Biochemistry22, 3763.
Del Mar, E.G., et al. 1980. Biochemistry19, 468.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C Largman
Biochemistry, 22(16), 3763-3770 (1983-08-02)
Proelastase has been purified to homogeneity from rat pancreatic tissue by a combination of CM-Sephadex and immobilized protease inhibitor affinity resins. Trypsin activation yields an elastolytic enzyme that possesses a specificity toward small hydrophobic residues in synthetic amide substrates, similar
E G Del Mar et al.
Biochemistry, 19(3), 468-472 (1980-02-05)
The substrate specificity of human pancreatic elastase 2 was investigated by using a series of peptide p-nitroanilides. The kinetic constants, kcat and Km, for the hydrolysis of these peptides revealed that this serine protease preferentially hydrolyzes peptides containing P1 amino

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service