Skip to Content
Merck
All Photos(1)

Key Documents

260920

Sigma-Aldrich

DNA Methyltransferase Inhibitor

The DNA Methyltransferase Inhibitor, also referenced under CAS 32675-71-1, controls the biological activity of DNA Methyltransferase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

DNA Methyltransferase Inhibitor, 2-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)-3-(1H-indol-3-yl)propionic acid, RG108, DNA MTase Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 199.00

CHF 199.00


Estimated to ship on30 May 2025



Select a Size

Change View
10 MG
CHF 199.00

About This Item

Empirical Formula (Hill Notation):
C19H14N2O4
CAS Number:
Molecular Weight:
334.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

CHF 199.00


Estimated to ship on30 May 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

potency

115 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL
methanol: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

[nH]1c2c(c(c1)CC(N3C(=O)c4c(cccc4)C3=O)C(=O)O)cccc2

InChI

1S/C19H14N2O4/c22-17-13-6-1-2-7-14(13)18(23)21(17)16(19(24)25)9-11-10-20-15-8-4-3-5-12(11)15/h1-8,10,16,20H,9H2,(H,24,25)

InChI key

HPTXLHAHLXOAKV-UHFFFAOYSA-N

General description

A cell-permeable, specific DNA methyltransferases inhibitor (IC50 = 115 nM for CpG methylase M.SssI) that displays anti-proliferative, but not cytotoxic, properties. Unlike the commonly used inhibitor 5-azacytidine, RG108 directly interacts with DNA methyltransferase active site via its carboxy group.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CPG methylase M.Sssl
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Brueckner, B., et al. 2005. Cancer Res.65, 6305.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service