Skip to Content
Merck
All Photos(1)

Key Documents

239836

Sigma-Aldrich

Cyclophilin A Inhibitor

The Cyclophilin A Inhibitor, also referenced under CAS 1186372-20-2, controls the biological activity of Cyclophilin A. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

Cyclophilin A Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 140.00

CHF 140.00


Estimated to ship on08 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
CHF 140.00

About This Item

Empirical Formula (Hill Notation):
C21H14ClFN2O2
CAS Number:
Molecular Weight:
380.80
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 140.00


Estimated to ship on08 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Fc1c(c(ccc1)Cl)C(=O)NC(=O)NC2c3c(cccc3)c4c2cccc4

InChI

1S/C21H14ClFN2O2/c22-16-10-5-11-17(23)18(16)20(26)25-21(27)24-19-14-8-3-1-6-12(14)13-7-2-4-9-15(13)19/h1-11,19H,(H2,24,25,26,27)

InChI key

SFNLLCUAISZNRV-UHFFFAOYSA-N

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuya Tsuda et al.
Journal of pharmacological sciences, 142(3), 109-115 (2019-12-29)
Arterial calcification, a risk factor of cardiovascular events, develops with differentiation of vascular smooth muscle cells (VSMCs) into osteoblast-like cells. Cyclophilin A (CypA) is a peptidyl-prolyl isomerase involved in cardiovascular diseases such as atherosclerosis and aortic aneurysms, and rho-associated protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service