Skip to Content
Merck
All Photos(1)

Key Documents

23-060-M

Sigma-Aldrich

IKKγ

Synonym(s):

NF-kappa-B essential modulator. Inhibitor of nuclear factor kappa-B kinase subunit gamma, FIP-3, NEMO, IKKg

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CHF 609.00
23-060M
CHF 3’520.00

CHF 609.00


Estimated to ship on14 May 2025



Select a Size

Change View
10 μG
CHF 609.00
23-060M
CHF 3’520.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

CHF 609.00


Estimated to ship on14 May 2025


biological source

human

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

GenBank accession no.

General description

IKKγ has been shown to play a critical role in NF-κB activation, as the regulatory subunit of IκB kinase, which phosphorylates inhibitors of NF-κB leading to their degradation. Upon cell stimulation, IKKγ can be modified by Lys-63-linked or linear polyubiquitination, both of which are suggested to play key roles in cell signalling, including NF-κB activation. One of the E3 ligases responsible for Lys-63-linked IKKγ polyubiquitination is TRAF6, which participates in several signalling pathways controlling immunity, skin development, osteoclast formation and brain functions.

Application

Ubiquitination Cascade Comoponent: Substrate

Physical form

Purified using glutathione sepharose.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service