Skip to Content
Merck
All Photos(1)

Key Documents

219379

Sigma-Aldrich

Cathepsin K Inhibitor II

The Cathepsin K Inhibitor II controls the biological activity of Cathepsin K. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Cathepsin K Inhibitor II, Inhibitor Boc-I, 1-(N-Benzyloxycarbonyl-leucyl)-5-(N-Boc-phenylalanyl-leucyl)carbohydrazide, Z-L-NHNHCONHNH-LF-Boc

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 152.00

CHF 152.00


Estimated to ship on08 May 2025



Select a Size

Change View
1 MG
CHF 152.00

About This Item

Empirical Formula (Hill Notation):
C35H51N7O8
Molecular Weight:
697.82
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 152.00


Estimated to ship on08 May 2025


Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable peptidyl bis-carbohydrazide compound that acts as a potent, selective, and reversible inhibitor of cathepsin K (Ki = 6 nM). Reported to completely block cathepsin K activity in primary human synovial fibroblast cultures at a concentration of 1 µM. Also shown to inhibit the activity of cathepsin B and papain at higher concentrations (Ki = 510 nM and 1.2 µM, respectively). Does not significantly inhibit cathepsin L.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
cathepsin K
Product does not compete with ATP.
Reversible: yes
Target Ki: 6 nM against cathepsin; 510 nM, 1.2 µM, against cathepsin B and papain, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Boc-Phe-Leu-NHNH-CO-NHNH-Leu-Z

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wang, D., et al. 2002. Biochemistry41, 8849.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service