Skip to Content
Merck
All Photos(1)

Key Documents

218763

Sigma-Aldrich

Caspase-6 Substrate III, Fluorogenic

A fluorogenic substrate for caspase-6 (Mch2).

Synonym(s):

Caspase-6 Substrate III, Fluorogenic, Z-VEID-AFC

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 247.00

CHF 247.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CHF 247.00

About This Item

Empirical Formula (Hill Notation):
C38H44F3N5O12
CAS Number:
Molecular Weight:
819.78
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 247.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

solubility

DMSO: 5 mg/mL

fluorescence

λex ~400 nm
λem ~505 nm

shipped in

ambient

storage temp.

−20°C

SMILES string

FC(F)(F)C1=CC(=O)Oc2c1ccc(c2)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)OCc3ccccc3)C(C)C)CCC(=O)O)[C@H](CC)C)CC(=O)O

InChI key

OEVAAZFKVRJQQH-UXAYAQJLSA-N

General description

A fluorogenic substrate for caspase-6 (Mch2). Reaction can be monitored visually or quantitatively by a blue to green shift in fluorescence upon cleavage of the AFC fluorophore (excitation maximum: ~400 nm; emission maximum: ~505 nm).

Biochem/physiol Actions

Cell permeable: no
Primary Target
A fluorogenic substrate for caspase-6 (Mch2
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Val-Glu-Ile-Asp-AFC

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service